missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin Stain Solution, Modified Harris Formulation (Mercury Free), Nuclear Stain for General Histology and Cytology, Ricca Chemical

$157.15 - $1,292.74

Chemical Identifiers

CAS 7732-18-5,7784-26-1,64-17-5,517-28-2,7758-05-6,7732-18-5 (Water, Deionized)
CAS Min % 89.0,5.32,3.55,0.44,0.05
CAS Max % 92.0,5.54,3.69,0.46,0.05
Physical Form Liquid
Chemical Name or Material Hematoxylin Stain Solution, Modified Harris Formulation (Mercury Free)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
3530-32 SDS Ricca Chemical Company
RI353032
Glass amber 1L Each for $157.15
3530-5 Ricca Chemical Company
RI35305
Cubitainer 20L Each for $1,292.74
Specifications

Description

Nuclear Stain for General Histology and Cytology
Description & Specifications

Chemical Identifiers

7732-18-5,7784-26-1,64-17-5,517-28-2,7758-05-6,7732-18-5 (Water, Deionized)
92.0,5.54,3.69,0.46,0.05
Hematoxylin Stain Solution, Modified Harris Formulation (Mercury Free)
89.0,5.32,3.55,0.44,0.05
Liquid

Specifications

7732-18-5,7784-26-1,64-17-5,517-28-2,7758-05-6,7732-18-5 (Water, Deionized)
92.0,5.54,3.69,0.46,0.05
Glass amber
Dark-purple
Hematoxylin Stain Solution, Modified Harris Formulation (Mercury Free)
89.0,5.32,3.55,0.44,0.05
Liquid
Mercury Free
1L