missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hematoxylin Hydrate MP Biomedicals
Supplier: Mp Biomedicals 0210192205
Specifications
Hematoxylin | |
517-28-2 | |
Natural Black I | |
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O | |
302.282 | |
356.4 |
Hydrate | |
C16H14O6 | |
WZUVPPKBWHMQCE-VYIIXAMBSA-N | |
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol | |
45029742 | |
5g |
Chemical Identifiers
517-28-2 | |
302.282 | |
Natural Black I | |
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol |
C16H14O6 | |
WZUVPPKBWHMQCE-VYIIXAMBSA-N | |
45029742 | |
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O |