missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hematoxylin hydrate, 80% (dry wt.), water <8%

Manufacturer:  Alfa Aesar™ 4722322

 View more versions of this product

Catalog No. AA4722322



Specifications

Specifications

Hematoxylin hydrate
Brown to Cream
100g
517-28-2
C16H14O6
91399
hematoxylin, hydroxybrasilin
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
302.282
302.29 (Anhydrous)
80%
Light Sensitive
~200°C (decomposition)
Water <8%
(dry weight)
MFCD00078111
14,4637
WZUVPPKBWHMQCE-VYIIXAMBSA-N
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
45029742
Powder

Chemical Identifiers

517-28-2
302.282
WZUVPPKBWHMQCE-VYIIXAMBSA-N
45029742
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
C16H14O6
MFCD00078111
hematoxylin, hydroxybrasilin
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : MH7875000

TSCA : TSCA

SDS Documents