missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin, Fisher BioReagents

$471.57 - $489.74

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.282
InChI Key WZUVPPKBWHMQCE-VYIIXAMBSA-N
Synonym Natural Black I
PubChem CID 45029742
IUPAC Name (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
SMILES C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP242425 SDS View Documents Fisher BioReagents
BP242425
25g Glass Bottle Each for $471.57
Case of 6 Each for $2,829.42
 
BP2424100 SDS View Documents Fisher BioReagents
BP2424100
100g Glass Bottle Each for $489.74
Case of 6 Each for $2,938.44
 
Description

Description

Hematoxylin is used as a stain in microscopy.
Specifications

Chemical Identifiers

517-28-2
302.282
Natural Black I
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
C16H14O6
WZUVPPKBWHMQCE-VYIIXAMBSA-N
45029742
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O

Specifications

517-28-2
C16H14O6
Natural Black I
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
302.282
302.28
Glass Bottle
100°C
25g
Pass Test
15, 4672
WZUVPPKBWHMQCE-VYIIXAMBSA-N
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
45029742
Powder/Solid
75290
Beige
Hematoxylin
Safety and Handling

Safety and Handling

Recommended Storage : RT