missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin, Fisher BioReagents

Manufacturer:  Fisher BioReagents BP242425

 View more versions of this product

Catalog No. BP242425




Description

Description

Hematoxylin is used as a stain in microscopy.
Specifications

Specifications

Hematoxylin
Pass Test
15, 4672
WZUVPPKBWHMQCE-VYIIXAMBSA-N
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
45029742
Powder/Solid
75290
Beige
517-28-2
C16H14O6
Natural Black I
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
302.282
302.28
Glass Bottle
100°C
25g

Chemical Identifiers

517-28-2
302.282
Natural Black I
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
C16H14O6
WZUVPPKBWHMQCE-VYIIXAMBSA-N
45029742
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents