missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin, MilliporeSigma™

Manufacturer:  Emd Millipore Corporation 23416

Catalog No. M23416



Specifications

Specifications

Hematoxylin
517-28-2
WZUVPPKBWHMQCE-VYIIXAMBSA-N
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
45029742
Certified Biological Stain
100g
H & E Stain, For Microscopy
C16H14O6
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
302.282
Liquid
Glass Bottle

Chemical Identifiers

517-28-2
302.282
45029742
C1C2=CC(=C(C=C2C3C1(COC4=C3C=CC(=C4O)O)O)O)O
C16H14O6
WZUVPPKBWHMQCE-VYIIXAMBSA-N
(6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
Safety and Handling

Safety and Handling

Recommended Storage : Store at +5°C to +3°C

Documents