Please login to your online account to display your discounted pricing

Hematoxylin, 85%, pure, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  411185000

 View more versions of this product

Catalog No. AC411185000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Loss on Drying 10 % max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
UV_Vis 80 % min.
Color Pale beige to brown
Quantity 500g
Chemical Name or Material Hematoxylin
CAS 517-28-2
Purity Grade Pure
Physical Form Pale beige to brown powder
Assay Percent Range 85%
Molecular Formula C16H14O6·xH2O
Formula Weight 302.27g/mol
MDL Number MFCD00078111
Merck Index 15, 4672
Synonym C.I. 75290
Health Hazard 1

Specifications

Chemical Name or Material Hematoxylin
CAS 517-28-2
Purity Grade Pure
Assay Percent Range 85%
Molecular Formula C16H14O6·xH2O
Formula Weight 302.27g/mol
MDL Number MFCD00078111
Synonym C.I. 75290

Indicator

Accessories

SDS & Safety Documents