Please login to your online account to display your discounted pricing

Hematoxylin, 85%, pure, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hematoxylin
CAS 517-28-2
Purity Grade Pure
Assay Percent Range 85%
Molecular Formula C16H14O6·xH2O
Formula Weight 302.27
MDL Number MFCD00078111
Synonym C.I. 75290
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC411181000

  SDS
thermo fisher scientific
411181000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC411180250

  SDS
fisher scientific acros
411180250
Glass Bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 302.27
Physical Form Powder
Quantity 100g
Color Pale beige to brown
Merck Index 15, 4672
Purity Grade Pure
Chemical Name or Material Hematoxylin
UV_Vis 80 % min.
Infrared Spectrum Authentic
Loss on Drying 10 % max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Synonym C.I. 75290
CAS 517-28-2
Molecular Formula C16H14O6·xH2O
Assay Percent Range 85%
MDL Number MFCD00078111
Health Hazard 1

Indicator