Please login to your online account to display your discounted pricing

Hematoxylin, 85%, pure, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  411181000

 View more versions of this product

Catalog No. AC411181000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 302.27
Physical Form Powder
Quantity 100g
Color Pale beige to brown
Merck Index 15, 4672
Purity Grade Pure
Chemical Name or Material Hematoxylin
UV_Vis 80 % min.
Infrared Spectrum Authentic
Loss on Drying 10 % max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Synonym C.I. 75290
CAS 517-28-2
Molecular Formula C16H14O6·xH2O
Assay Percent Range 85%
MDL Number MFCD00078111
Health Hazard 1

Specifications

Chemical Name or Material Hematoxylin
CAS 517-28-2
Purity Grade Pure
Assay Percent Range 85%
Molecular Formula C16H14O6·xH2O
Formula Weight 302.27
MDL Number MFCD00078111
Synonym C.I. 75290
Accessories

SDS & Safety Documents