Please login to your online account to display your discounted pricing

Alfa Aesar™ Hematoporphyrin dihydrochloride

$61.78 - $61.78

Chemical Identifiers

Chemical Name or Material Hematoporphyrin dihydrochloride
CAS 17696-69-4
Molecular Formula C34H38N4O6·2HCl
Formula Weight 671.62
MDL Number MFCD00013470
Beilstein 78957
Merck Index 14,4636
Synonym Haematoporphyrin dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1857903

  SDS
alfa aesar™
A1857903
1g Each for $61.78
Description & Specifications

Specifications

Formula Weight 671.62
Chemical Name or Material Hematoporphyrin dihydrochloride
TSCA TSCA
Molecular Formula C34H38N4O6·2HCl
MDL Number MFCD00013470
CAS 17696-69-4
Beilstein 78957
Merck Index 14,4636
Synonym Haematoporphyrin dihydrochloride
RTECS Number TS5505000
Quantity 1g

Applications
Endogenous porphyrin formed by the acid hydrolysis of hemoglobin. It is employed as an active pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in pyridine (5%).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.