Please login to your online account to display your discounted pricing

Alfa Aesar™ Hematin, 97%

$584.27 - $584.27

Chemical Identifiers

Chemical Name or Material Hematin
CAS 15489-90-4
Molecular Formula C34H33FeN4O5
Formula Weight 633.51g/mol
MDL Number MFCD00011615
Merck Index 14,4635
Synonym Haematin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1851814

 
alfa aesar™
A18518-14
25g Each for $584.27
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Hematin
CAS 15489-90-4
Assay 97%
Molecular Formula C34H33FeN4O5
Formula Weight 633.51g/mol
MDL Number MFCD00011615
Merck Index 14,4635
Synonym Haematin
Recommended Storage 2° to 8°C
RTECS Number NO6725000