missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Helios Armenian Hamster, PE-Cy7, Clone: 22F6, eBioscience™

Armenian Hamster Monoclonal Antibody

$187.44 - $469.92

Specifications

Antigen Helios
Clone 22F6
Host Species Armenian Hamster
Gene Alias Ikaros family zinc finger 2, IKZF2
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123470 Life Technologies
LS25988341
25 Tests Each for $187.44
501123471 Life Technologies
LS25988342
100 Tests Each for $469.92
Specifications

Description

This 22F6 monoclonal antibody reacts with human, mouse and canine Helios. Helios is a zinc-finger transcription factor first identified for its similarity to Ikaros. It is expressed in hematopoietic stem cells in both the embryo and adult mice. While the highest expression is found in regulatory T cells, Helios is not expressed in mature B cells, dendritic cells or myeloid cells. Helios can form a homodimer as well as heterodimerize with other Ikaros family members to regulate transcription. Eight isoforms of Helios exist, some of which possess a dominant-negative function that has also been found to be upregulated in lymphoid malignancies, including T-acute lymphoblastic leukemia.

22F6 has been reported to cross react with rat Helios.
Description & Specifications

Specifications

Helios
Armenian Hamster
Flow Cytometry (Intracellular Staining)
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.015μg)/test
22F6
Ikaros family zinc finger 2, IKZF2
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer