Please login to your online account to display your discounted pricing

Alfa Aesar™ Helicin hydrate

$31.86 - $80.50

Chemical Identifiers

Chemical Name or Material Helicin hydrate
CAS 618-65-5
Molecular Formula C13H16O7·xH2O
Formula Weight 284.27 (Anhydrous)
MDL Number MFCD00006589
Beilstein 89596
Merck Index 14,4628
Synonym Salicylaldehyde-beta-D-glucopyranoside hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1283003

 
alfa aesar™
L1283003
1g Each for $80.50

AAL12830MD

 
alfa aesar™
L12830MD
250mg Each for $31.86
Description & Specifications

Specifications

Formula Weight 284.27 (Anhydrous)
Chemical Name or Material Helicin hydrate
Molecular Formula C13H16O7·xH2O
MDL Number MFCD00006589
CAS 618-65-5
Synonym Salicylaldehyde-beta-D-glucopyranoside hydrate
Beilstein 89596
Merck Index 14,4628
Optical Rotation −57° (c=1.5 in Water)
Quantity 1g