Please login to your online account to display your discounted pricing
Heidolph™ Tuttnauer™ Tabletop Autoclaves Accessories
Accessories and spare parts for Heidolph Tuttnauer Autoclaves
$81.22 - $1,543.13
For Use With (Equipment) Tuttnauer benchtop sterilizer
View More Specs
Catalog Number Mfr. No. Model Price Quantity    
heidolph north america
2340EP, 2540EP, 2540EPK, 3850EP, 3870EP Pack of 5 Each for $81.22
heidolph north america
3870M, 3870E, 3870EP Each for $1,097.07
heidolph north america
Any autoclave Pack of 10 Each for $180.98
heidolph north america
2340E, 2540E, 3850E, 3870E Each for $1,543.13
Description & Specifications
For Use With (Equipment) Tuttnauer benchtop sterilizer
Chamber BRITE Sterilizer Cleaner
  • Fast, convenient and effective way to clean & descales steam sterilizers, including the chamber, reservoir & tubing
  • Convenient premeasured packets replace messy liquids, measuring and scrubbing
  • Simply sprinkle one packet into the chamber and run. Recommended every 20 cycles of use or once per week, depending on usage
  • Regular use optimizes autoclave function & extends operating life
Printer Paper
  • Package of 5 rolls, for EP modelsOther Accessories
    • Replacement Door Gasket
    • Stand for Tuttnauer Models 3850 and 3870
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.