Please login to your online account to display your discounted pricing
Heidolph™ Radleys Heat-On Multi-Well Packages
Replace messy oil baths and heating mantles; avoid spills and make chemistry safer, cleaner and faster
$1,399.78 - $1,818.24
Specifications
Closure Material Aluminum
For Use With (Equipment) Heat-On multi-well magnetic stirrer
Material Aluminum
View More Specs
Includes:  Multi-Well holder (1), inserts for 25, 50 and 100mL flasks, and adaptor for 135mm hotplates
Products
Catalog Number Mfr. No. Coating Includes Price Quantity    
05-404-154
heidolph north america
015852020CA
Anodized Two 25mL, two 50mL and two 100mL inserts, adapter plate for 135mm hotplates Each for $1,818.24
05-404-153
heidolph north america
015852010CA
Fluoropolymer 25, 50 and 100mL insert Each for $1,399.78
Description & Specifications
Specifications
Closure Material Aluminum
For Use With (Equipment) Heat-On multi-well magnetic stirrer
Material Aluminum
Not all block designs are the same; test results show that Heat-On heats up 66% faster and uses 30% less energy than other leading brands of blocks.
  • No oil baths or heating mantles required- safer operation with less clean-up
  • Can be used with all common hotplates up to 145mm in diameter; adaptor for 135mm hotplates included in Heat-On system packages
  • Heats up to 2000°C, and even up to 2600°C for short periods
  • Solid aluminum for fast and even heat transfer from the hotplate into the sample
  • Lightweight and deep well design for increased surface contact contribute to fast heat transfer with added safety — no sticking or broken flasks to worry about
  • Fluoropolymer coating for superior chemical resistance; optional anodized coating also available
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.