Heidolph™ Platform Shaker Accessories
Separatory funnel clamps for Heidolph Platform Shakers as indicated
Supplier:  Heidolph™ 036303450
 View more versions of this product
Catalog No. 13-889-845
Description
Description
  • Choice of clamp either for volumes 250, 500, and 1000 mL or 2000 mL separatory funnel
Specifications
Specifications
250, 500 and 1000 mL
Separatory funnel clamp
Polymax 2040, Promax 2020, Unimax 2010 platform shakers