Heidolph™ Hei-VAP Industrial and Laborota 20 Rotary Evaporator Accessories
Accessories for use with Heidolph Hei-VAP Industrial and Laborota 20 rotary evaporators only
Supplier: Heidolph™ 036303080
Description
- The vacuum sensor controls and displays the actual vacuum controls in addition the maximum permissible pressure.
- The vacuum valve controls vacuum.
- Vapor temperature sensor measure the temperature of vapors in the condenser.
- Tubing set is 6 meter of vacuum and 10 meter of enforced cooling tube including hose clamps.
Specifications
Vapor Temperature Sensor | |
Hei-VAP Industrial and Laborota 20 Rotary Evaporator |