Heidolph™ Laborota™ 20 Distimatic Rotary Evaporators
For unattended and continuous operation including automatic release of residue, automatic module is compatible with all Labrota 20 models
Manufacturer:  Heidolph Instruments 036064700CA
 View more versions of this product
Catalog No. 13-889-372
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Unattended and continuous operation including automatic release of residue
Distimatic Rotary Evaporator
Automatic release of residue and adapter piece of Glassware set R, sensor
Documents