Heidolph™ Julabo F250 Recirculating Cooler
Environmentally-friendly operation with low energy consumption
Manufacturer:  Heidolph™ 048100068CA
Catalog No. 13-889-536
Includes: Two each barbed fittings for tubing 8 and 10mm inner dia. (pump connections M10 x 1 male)
Description
Description
  • Cooler with compact design and filling level indicator
  • Equipped with straightforward filling and draining
  • Splash-proof membrane keypad with LED temperature display
  • Used with water, water/glycol, JULABO Thermal G
  • Needs very little space
For basic cooling tasks
Specifications
Specifications
115 V 60 Hz
0.35 bar
M10 x 1
±0.5°C
-10°C to +40°C
15L/min.
F250
Air
27 kg
Documents