Please login to your online account to display your discounted pricing
Heidolph™ Immersion Blenders - SilentCrusher M Accessories - Generators
Generators used with Heidolph SilentCrusher M blender
$2,242.77 - $2,572.95
Specifications
Type Generator
For Use With (Application) Ideal for applications under vacuum down to 20mbar or pressure up to 6bar
For Use With (Equipment) SilentCrusher M blender
Material Stainless Steel
View More Specs
Products
Catalog Number Mfr. No. Model Noise Level Particle Size (Metric) Speed Price Quantity    
13-900-806
heidolph north america
036300840CA
22 G 73dBA <0.04cm 20000rpm Each for $2,572.95
13-900-801
heidolph north america
036300790CA*
18 F 74dBA <0.04cm 26000rpm Each for $2,242.77
Description & Specifications
Specifications
Type Generator
For Use With (Application) Ideal for applications under vacuum down to 20mbar or pressure up to 6bar
For Use With (Equipment) SilentCrusher M blender
Material Stainless Steel
  • High quality generators made of corrosion-resistant stainless-steel AISI 316
  • Easily disassembled for quick and convenient cleaning making them ideal for multiple samples
  • Autoclavable up to 130°C or air sterlization up to 180°C
  • Designed with interchangeable ball bearings and chemical-resistant PTFE for years of usage
For larger samples it may be necessary to pre-crush samples. In this case, generators which feature additional sprockes are recommended (except for 18 G, 18 DG, and 22 G). For applications under vacuum down to 20 mbar or pressure up to 6 bar, sealed generators are recommended (8 DF, 12 DF, 12 DG, 18 DF, and 18 DG).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.