Heidolph™ Heat-On Blocks
For use with Heidolph MR Hei Series magnetic stirrers
Manufacturer:  Heidolph™ 015882080CA
 View more versions of this product
Catalog No. 05-404-164
Description
Description
Each Heat-On block is a standalone product that can be placed directly onto the stirring hot plate.
  • Fit precisely to every flask ensuring maximum surface contact and fast heat transfer
  • Unique design prevents glass breakage, protecting user from fires and contamination due to spilled chemicals or solvents
  • High temperature range provides unlimited safety for applications up to 260degreeC
  • Unique PTFE coating allows for superior chemical resistance - providing many years of service life
  • Provides precise temperature control
  • 1000mL of water heats up to 66% faster than conventional oil baths
  • 150mL of water boils in less than 11 minutes
Specifications
Specifications
33.8 oz.
Fluoropolymer
Aluminum
1000 mL
MR Hei Series Magnetic Stirrer
Heat Block
Documents