Please login to your online account to display your discounted pricing
Heidolph™ Cool-It Cooling Bowl and Hotplate Adaptor
For use with Heidolph stirring hotplates
$890.08 - $890.08
Specifications
For Use With (Equipment) Heidolph stirrers
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
13-900-173
heidolph north america
015882420CA
Small Bowl and Lid Each for $890.08
Description & Specifications
Specifications
For Use With (Equipment) Heidolph stirrers
Compact, affordable cooling and stirring work station designed to accommodate single round bottom flasks from 10mL to 2L.
  • Designed to fit on a stirring hotplate for controlled stirring through a magnetic stirring bar in the reaction flask
  • Cooling is obtained using an appropriate mixture such as dry ice and acetone
  • Cools down to -70C while stirring
  • Manufactured fro a robust HDPE outer with an encapsulated insulating foam core, creating an almost unbreakabloe product
  • Insulation minimizes condensation on outer surface, protecting the stirring hotplate
  • Unique non-drip pouring spout allows liquid to be easily drained without spilling or splashing solvents
  • Integral handle provides additional support for pouring
  • Lid ehnances cooling performance and reduces condensation and ice formation on the flask
Cool-It Bowls come complete with fitted lid. Adaptor plate for 135mm hotplate sold separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.