missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heidolph™ 2840 ELV/ELVC Tuttnauer™ Vertical Autoclaves

Top loading autoclaves with advanced microprocessor control panel and chamber made of 316Ti stainless steel

Manufacturer:  Heidolph North America 023210916CA

 View more versions of this product

Catalog No. 13-889-563

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Advanced Control system

  • F0 software control protects liquid media by minimisng exposire to high temperatures during sterilization
  • PID (Proportional Integral Differential) pressure control
  • Stores last 200 cycles in built-in memory
  • Up to 6 temperature sensors and 4 pressure sensors can be connected
  • 30 Identification codes and passwords for access level control
  • Controller and software comply with the 21 CFR part 11 standard
  • In/Out test (enables technical to check each system component separately)
  • Sterilization Temperature range 105° to 137°C
  • Optional Disinfection/Isothermal Temperature Range 60° to 105°C
  • Optional Independent Recording for cross-checking cycle measurements
  • Filter replacement notifications based on number of cycles
  • Optional Fast Liquid Cooling- (ELVC Models)
Multi-Color Display
  • Enables easy reading
  • Color indicates cycle stage
  • Built-in view of historical cycle data
  • Graphical display of Temperature and Pressure
  • Gives quick access to important information
  • Easily operated in mulitple languages

Applications:

For laboratories in research institutes, universities, pharmaceutical, food, chemical and biotechnological industries

Specifications

Specifications

Tuttnauer™ Vertical Autoclave
50/60Hz
Documents