missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Heavy Metals, plasma standard solution, Specpure™, Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL

Manufacturer:  Alfa Aesar™ 14657AE

 View more versions of this product

Catalog No. AA14657AE



Specifications

Specifications

Heavy Metals, plasma standard solution
Specpure™
MFCD02091713
Liquid
Ag, As, Ba, Cd, Cr, Hg, Pb, Se @ 100μg/mL
Matrix: 5% HNO3
UN3264
100mL
Documents