missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heavy-Duty Utility Cart

Handles large loads with ease

Manufacturer:  Fisher Science Education 452088BEI

 View more versions of this product

Catalog No. S68057



Description

Description

  • Load capacity: 500 lb. (227kg)
  • Sturdy, structural foam construction won't rust, dent, chip or peel
  • Rounded corners protect walls and furniture
  • Lightweight and maneuverable
  • Nonmarking 5 in. dia. casters (two fixed, two swivel)
  • Two sizes available

Order Info

Small cart L x W x H: 39 x 17.88 x 33.25 in. (99 x 45.4 x 84.5cm). Large cart L x W x H: 45.25 x 25.88 x 33.25 in. (115 x 65.7 x 84.5cm). Optional middle shelf available for both sizes.

Specifications

Specifications

Heavy-duty Utility Cart
45.25 x 25.88 x 33.25 in. (115 x 65.7 x 84.5 cm)
33.25 in.
Ergonomically push handle, molded-in customizable fittings, 2 lipped tray-shelf, rounded corners, 4 in. large nonmarking 2 fixed and 2 swivel casters, molded-in storage compartments, holsters, hooks and notches
115 cm
227 kg
2
25.88 in.
Beige
Transporting materials, supplies and heavy loads in almost any environment
84.5 cm
45.25 in.
500 lb.
Polypropylene
Medium
65.7 cm
SDS
Documents
Product Certifications
Promotions

Promotions