Please login to your online account to display your discounted pricing
Ansell™ Heavy-Duty Rubber Gloves
Long-wearing, natural rubber gloves with cotton lining.
Manufacturer: ansell™  113946D
 View more versions of this product
 Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. S47369
  • / Dozen of 12 Pair
Description & Specifications
Specifications
Size 10
Material Rubber, natural
Thickness (English) 18mil
Color Orange
Length (English) 13 in.
Length (Metric) 33cm
Certifications 21 CFR 177-2600, FDA
Exterior Finish Recessed diamond
For Use With (Application) canning and food processing, Detergents and cleaning liquids, handling Acids, Alkalies, Salts and Ketones, janitorial and maintenance work
Glove Cuff Style Straight
Lining Cotton Flock