Please login to your online account to display your discounted pricing
Fisher Scientific™ Heating Thermal Mixer II PROMO
For precise sample preparation when working with DNA, RNA, plasmids, cultures and enzymatic reactions
$5,299.70 - $5,299.70
Specifications
Type Thermal Mixer
Certifications/Compliance UL, CE, CSA approved
Speed 250-3000rpm, in increments of 50rpm
Timer 0 to 99 hours: 59 minutes: 59 seconds/hold
Depth (Metric) 27.94cm
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
11-700-176
fisher scientific™
BKL270900FPROMO
Conical Tube Blocks (15-600-333 and 15-600-332) Each for $5,299.70
11-700-177
fisher scientific™
BKL270900FPROMO11
Conical Tube Blocks (15-600-333) 2 Each Each for $5,299.70
11-700-178
fisher scientific™
BKL270900FPROMO12
Conical Tube Blocks (15-600-333 and 15-600-334) Each for $5,299.70
11-700-179
fisher scientific™
BKL270900FPROMO13
Conical Tube Blocks (15-600-333 and 15-600-335) Each for $5,299.70
11-700-180
fisher scientific™
BKL270900FPROMO14
Conical Tube Blocks (15-600-333 and 15-600-336) Each for $5,299.70
11-700-181
fisher scientific™
BKL270900FPROMO15
Conical Tube Blocks (15-600-333 and 15-600-337) Each for $5,299.70
11-700-182
fisher scientific™
BKL270900FPROMO16
Conical Tube Blocks (15-600-333 and 15-600-338) Each for $5,299.70
11-700-183
fisher scientific™
BKL270900FPROMO17
Conical Tube Blocks (15-600-333 and 15-600-339) Each for $5,299.70
11-700-184
fisher scientific™
BKL270900FPROMO18
Conical Tube Block (15-600-333) and Standard/Deep Microplate Block (15-600-340) Each for $5,299.70
11-700-185
fisher scientific™
BKL270900FPROMO19
Conical Tube Block (15-600-333) and PCR Plate Block (15-600-341) Each for $5,299.70
11-700-186
fisher scientific™
BKL270900FPROMO20
Conical Tube Block (15-600-333) and PCR Plate Block (15-600-342) Each for $5,299.70
Description & Specifications
Specifications
Type Thermal Mixer
Certifications/Compliance UL, CE, CSA approved
Speed 250-3000rpm, in increments of 50rpm
Timer 0 to 99 hours: 59 minutes: 59 seconds/hold
Depth (Metric) 27.94cm
Width (Metric) 17.78cm
Height (Metric) Platform 19.05cm
Electrical Requirements 12VDC via 115/230VAC adapter
Temperature Range (Metric) 15°C below ambient to 100°C
Operating Temperature (Metric) 10° to 30°C
Warranty Two years
  • Large color touch screen
  • Interchangeable blocks
  • Auto-recognition of blocks
  • Mixing and temperature control for small samples
  • For laboratory use
  • Designed by scientists for scientists
  • Easy to use and program
Intuitive menu navigation
  • Home display indicates step number and program name
  • Unit operating status is indicated by bright red text
  • “Stop, Play, Pause, Skip and Quick Mix” icons are easy to use
  • Instantly change parameters for one step
  • Turn off or on parameters as needed during process
  • Minimum and maximum parameters are automatically determined by block type
  • View and modify all steps on program screen
  • Automatically repeat one sequence as many times as needed
  • Use as many as steps as needed, up to five available
  • Easily select, launch, change, and save programs
  • Programs can have long text names for easy process recall
  • Turn on parameter lock to protect important processes
  • Stores up to 25 programs