Please login to your online account to display your discounted pricing

Fisherbrand™ DuraPorter™ Transport Box


Hold and store lab equipment and samples safely. Fisherbrand™ DuraPorter™ Transport Box is a durable, leak-proof container that is easy to load and locks securely. Accommodates a 72-place OneRack™ rack or features separators to create custom compartments. Available in three translucent colors.

Manufacturer: fisherbrand™  HS120052

 View more versions of this product

Catalog No. 12-006-950

  • / Each
 

Description & Specifications

Specifications

Quantity 1Each
Dimensions (L x W x H) 38.73 x 20.95 x 17.14cm
Color Clear
Width (Metric) 20.95cm
Length (Metric) 38.73cm
For Use With (Application) Transporting
For Use With (Equipment) 13 and 16mm tubes in a 72-place rack
Includes Separators, three lid clasps, water-tight seal
Material Polycarbonate
Product Type Transport Box
Height (Metric) 17.14cm

Designed for secure, convenient and customizable storage

  • Sturdy translucent polycarbonate provides content visibility and is fully autoclavable
  • Available in three colors (clear, translucent yellow or translucent red) for easy user/owner identification
  • Silicone water-tight seal and three lid clasps provide secure closure
  • Carry handle folds neatly into body space allowing the units to be stacked
  • Lid opens a full 180°, allowing total access to contents and making it easier to clean
  • Will accommodate 13 and 16mm tubes in a 72-place OneRack™ or similar size/brand of rack
  • Use the separators (provided) to create your own compartment sizes, or use empty to transport larger products and equipment
Ideal for a variety of laboratory equipment and biological and clinical samples
  • Hazardous/sensitive research samples
  • Clinical and Phlebotomy samples
  • Blood bags and/or tissue/organ samples
  • Public Health samples
  • Laboratory instruments
  • Biotransport carrier