Please login to your online account to display your discounted pricing

Heat Transfer Kit


Observe and measure the transfer of heat between two water samples of different temperatures.

Manufacturer: hubbard scientific co  HUB3120

 View more versions of this product

Catalog No. S53012B

  • / Pack of 15 Each
 

Description & Specifications

Specifications

Includes Two insulated containers with slotted lids, a u-shaped aluminum transfer bar and two thermometers
For Use With (Application) To observe and measure the transfer of heat between two water samples of different temperatures
Product Type Heat Transfer Glass
Grade Level 6 to 12
Grades 6 to 12