Please login to your online account to display your discounted pricing

Heat Transfer Kit

Observe and measure the transfer of heat between two water samples of different temperatures.

$778.83 - $778.83

Specifications

For Use With (Application) To observe and measure the transfer of heat between two water samples of different temperatures
Grade Level 6 to 12
Includes Two insulated containers with slotted lids, a u-shaped aluminum transfer bar and two thermometers
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S53012

 
hubbard scientific co
HUB3122
Heat Transfer Kit CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S53012B

 
hubbard scientific co
HUB3120
Heat Transfer Glass Pack of 15 Each for $778.83
Description & Specifications

Specifications

For Use With (Application) To observe and measure the transfer of heat between two water samples of different temperatures
Grade Level 6 to 12
Includes Two insulated containers with slotted lids, a u-shaped aluminum transfer bar and two thermometers

  • Includes two insulated containers with slotted lids, a U-shaped aluminum transfer bar and two thermometers
  • Grades 6 to 12