Please login to your online account to display your discounted pricing

Heat Shock Protein Mouse anti-Human, Clone: STRSN, eBioscience™

Mouse Monoclonal Antibody

$131.95 - $317.55

Specifications

Antigen Heat Shock Protein
Clone STRSN
Host Species Mouse
Gene Alias heat shock protein beta-1, HSPB1, Hsp27
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122762

 
life technologies
LS14911280
25μg Each for $131.95

501122763

 
life technologies
LS14911282
100μg Each for $317.55
Description & Specifications

Specifications

Antigen Heat Shock Protein
Clone STRSN
Host Species Mouse
Gene Alias heat shock protein beta-1, HSPB1, Hsp27
Species Reactivity Human
Applications Enzyme Linked Immunosorbent Assay
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody STRSN recognizes human heat shock protein 27 (HSP27). The heat shock proteins are a large family of proteins that are induced in response to stressors such as changing temperatures, cytokine levels, and presence of various chemicals. Heat shock proteins serve as molecular chaperones, binding to other proteins to maintain proper protein structure or by folding denatured proteins and preventing activation of caspases that could lead to apoptosis. Unlike the larger HSP proteins, HSP27 belongs to a unique subgroup that is involved in apoptosis regulation. Expression of HSP27 is constitutive, but upon stress HSP27 is upregulated and can relocate from the cytoplasm to the nucleus. Increased HSP27 expression with certain carcinomas has been observed. In breast cancer, HSP27 expression has been shown to be responsive to estrogen levels. In both breast cancer and melanoma, high levels of HSP27 correlate with more aggressive tumors and decreased survival. Increased levels of HSP27 in the serum of patients with Chronic Obstructive Pulmonary Disease (COPD), breast cancer and melanoma may serve as a potential diagnostic marker. No crossreactivity is observed with HSP70, 90, 104 or 110.