Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heat Shock Protein 90β /hsp90β (HSP84) Ab-1, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

$274.69 - $1,221.61

Specifications

Antigen Heat Shock Protein 90beta/hsp90beta (HSP84) Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

RB118P

 
thermo scientific™ lab vision™
RB118P
Purified Ab with BSA and Azide 1mL Each for $1,129.80

RB118P0

 
thermo scientific™ lab vision™
RB118P0
Purified Ab with BSA and Azide 100µL Each for $274.69

RB118P1

 
thermo scientific™ lab vision™
RB118P1
Purified Ab with BSA and Azide 500µL Each for $755.16

RB118P1ABX

 
thermo scientific™ lab vision™
RB118P1ABX
Ab without BSA/Azide (Frozen) 500µL Each for $830.47

RB118PABX

 
thermo scientific™ lab vision™
RB118PABX
Ab without BSA/Azide (Frozen) 1mL Each for $1,221.61
Description & Specifications

Specifications

Antigen Heat Shock Protein 90beta/hsp90beta (HSP84) Ab-1
Concentration 1mg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Stress Response
Type Primary
Immunogen A synthetic peptiude corresponding to aa 2-13 (PEEVHHGEEEVE-Cys) from the N-terminus end of mouse heat shock protein 90Beta
Isotype IgG

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Heat Shock Protein 90β/hsp90β (HSP84) Ab-1, Rabbit Polyclonal Antibody.

Cells respond to heat and chemical stress by inducing the synthesis of a group of proteins referred to as heat shock proteins. HSP90 is present in cells at high levels in the cytoplasm and is composed of two separate gene products, HSP90α and HSP90β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 2-13

Immunogen: A synthetic peptide corresponding to aa 2-13 (PEEVHHGEEEVE-Cys) from the N-terminus of mouse heat shock protein 90β

Molecular Weight: 84 kDa

Positive Control: MAD109 cells or breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO