Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heat Shock Protein 90β /hsp90β (HSP84) Ab-1, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

Manufacturer: thermo scientific™ lab vision™  RB118P1ABX

 View more versions of this product

Catalog No. RB118P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG
Applications Western Blotting
Format Concentrated
Quantity 500µL
Regulatory Status RUO
Immunogen A synthetic peptiude corresponding to aa 2-13 (PEEVHHGEEEVE-Cys) from the N-terminus end of mouse heat shock protein 90Beta
Antigen Heat Shock Protein 90beta/hsp90beta (HSP84) Ab-1
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Concentration 1mg/mL
Classification Polyclonal
Research Discipline Stress Response
Type Primary

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments with Thermo Scientific™ Heat Shock Protein 90β/hsp90β (HSP84) Ab-1, Rabbit Polyclonal Antibody.

Cells respond to heat and chemical stress by inducing the synthesis of a group of proteins referred to as heat shock proteins. HSP90 is present in cells at high levels in the cytoplasm and is composed of two separate gene products, HSP90α and HSP90β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 2-13

Immunogen: A synthetic peptide corresponding to aa 2-13 (PEEVHHGEEEVE-Cys) from the N-terminus of mouse heat shock protein 90β

Molecular Weight: 84 kDa

Positive Control: MAD109 cells or breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO