Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heat Shock Protein 60 (HSP60) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and electron microscopy experiments.

Manufacturer: thermo scientific™ lab vision™  MS120P1ABX

 View more versions of this product

Catalog No. MS120P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Heat Shock Protein 60 (HSP60) Ab-1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone LK1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Recombinant HSP60 protein
Isotype IgG1
Quantity 100μL
Regulatory Status RUO
Research Discipline Stress Response
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Chicken
Target Species Hamster
Target Species Human
Target Species Murine
Target Species Rat

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and electron microscopy experiments with Thermo Scientific™ Heat Shock Protein 60 (HSP60) Ab-1, Mouse Monoclonal Antibody.

A wide variety of environmental and pathophysiological stressful conditions trigger the synthesis of heat shock proteins (hsp's) or stress response proteins (srp's). hsp60 is a potential antigen in a number of autoimmune diseases including human arthritis.

Host Species: Mouse

Clone: LK1

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Hamster and Chicken. Does not react with bacteria, helminths and spinach. Others not known.

Epitope: aa 383-447

Immunogen: Recombinant HSP60 protein

Molecular Weight: 60 kDa

Positive Control: BT474 or MAD109 cells, Breast CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO