missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heat Shock Protein 27 (HSP27) Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.

$278.45 - $1,449.92

Specifications

Antigen Heat Shock Protein 27 (HSP27) Ab-1
Clone G3.1
Applications Western Blot
Classification Monoclonal
Host Species Murine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS101B Thermo Scientific™ Lab Vision™
MS101B
Biotinylated Ab with BSA and Azide 1mL Each for $1,449.92
MS101B0 Thermo Scientific™ Lab Vision™
MS101B0
Biotinylated Ab with BSA and Azide 100μL Each for $368.73
MS101B1 Thermo Scientific
MS101B1
Biotinylated Ab with BSA and Azide 500μL Each for $910.20
MS101P Thermo Scientific™ Lab Vision™
MS101P
Purified Ab with BSA and Azide 1mL Each for $1,131.33
MS101P0 Thermo Scientific™ Lab Vision™
MS101P0
Purified Ab with BSA and Azide 100μL Each for $278.45
MS101P1 Thermo Scientific™ Lab Vision™
MS101P1
Purified Ab with BSA and Azide 500μL Each for $701.67
MS101P1ABX Thermo Scientific™ Lab Vision™
MS101P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $777.37
MS101PABX Thermo Scientific™ Lab Vision™
MS101PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,186.90
MS101R7 Thermo Scientific™ Lab Vision™
MS101R7
Ready-to-use for IHC (UVLP) 7mL Each for $666.71
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Heat Shock Protein 27 (HSP27) Ab-1, Mouse Monoclonal Antibody.

Hsp27 is identical to the estrogen-induced“p29” and“24K” protein. About 50% of breast carcinomas are positive for hsp27 especially those that are also positive for estrogen and/or progesterone receptor. HSP27 has also been implicated in drug resistance in cancer cells.

Host Species: Mouse

Clone: G3.1

Isotype: IgG1

Species Reactivity: Human, Chimpanzee and Monkey. Does not react with mouse and rat. Others not known.

Epitope: Not determined

Immunogen: Partially purified HSP27 (earlier called 24K) protein from breast cancer MCF-7 cells

Molecular Weight: 24-27 kDa

Positive Control: BT474 or MDA-MB-453 cells, Breast CA

Cellular Localization: Cytoplasmic/nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Heat Shock Protein 27 (HSP27) Ab-1
Western Blot
Murine
Stress Response
Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells
G3.1
Monoclonal
RUO
Primary
IgG1