Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Heat Shock Factor 1 (HSF1) Ab-1, Rat Monoclonal Antibody

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments.

$272.06 - $1,209.59

Specifications

Antigen Heat Shock Factor 1 (HSF1) Ab-1
Clone 4B4
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

RT405P

 
thermo scientific™ lab vision™
RT405P
200µg/mL 1mL Each for $1,112.20

RT405P0

 
thermo scientific™ lab vision™
RT405P0
200µg/mL 100µL Each for $272.06

RT405P1

 
thermo scientific™ lab vision™
RT405P1
200µg/mL 500µL Each for $730.18

RT405P1ABX

 
thermo scientific™ lab vision™
RT405P1ABX
1mg/mL 100µL Each for $826.45

RT405PABX

 
thermo scientific™ lab vision™
RT405PABX
1mg/mL 200µL Each for $1,209.59
Description & Specifications

Specifications

Antigen Heat Shock Factor 1 (HSF1) Ab-1
Clone 4B4
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Format Concentrated
Host Species Rat
Regulatory Status RUO
Research Discipline Stress Response
Type Primary
Immunogen Recombinant mouse HSF1 protein (aa1-503)
Isotype IgG1

Ensure accurate, reproducible results in western blotting, immunofluorescence, immunoprecipitation and gel supershift experiments with Thermo Scientific™ Heat Shock Factor 1 (HSF1) Ab-1, Rat Monoclonal Antibody.

HSF1 responds to cellular stress signals such as heat, heavy metals, and oxidative reagents; binds to heat shock response elements (HSEs) in the upstream region of heat shock gene and activates their transcription. Some laboratories have reported that the constitutive, non-DNA binding, monomeric form of inactive HSF1 is located in the cytoplasm. Upon activation, HSF1 is trimerized and is localized in nucleus where it binds DNA.

Host Species: Rat

Clone: 4B4

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 425-439 of mouse HSF1

Immunogen: Recombinant mouse HSF1 protein (aa 1-503)

Molecular Weight: 70-85 kDa (depending upon the source and state of cells)

Positive Control: LS174T or MAD109 cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO