missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Heat Inactivated, Fetal Bovine Serum, Qualified, ONE SHOT™, USDA Approved Regions DFS Item

Heat-inactivated serum in a One-Shot 50-mL bottle increases workflow benefits, while maintaining serum quality. The 50-mL bottle eliminates the need to aliquot and reduces the risk of contamination.

$103.00 - $968.00

Specifications

Origin USDA Approved Regions
Grade Qualified
Heat Inactivated Yes
Format One Shot
Shipping Considerations Dry Ice
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A3840202 View Documents Gibco™
LSA3840202
10 x 50mL Each for $968.00
 
A3840201 View Documents Gibco™
LSA3840201
50mL Each for $103.00
 
Description

Description

One-Shot 50-mL bottle features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500-mL bottles
Specifications

Specifications

USDA Approved Regions
Yes
Dry Ice
≤50 EU/mL
Sterile-Filtered
Qualified
One Shot
−10°C
≤25 mg/dL