missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HEAT hydrochloride, Tocris

Highly selective α1 antagonist

Manufacturer:  R & D Systems RDS053510

 View more versions of this product

Catalog No. 053510


 

Specifications Description & Specifications

Specifications

HEAT hydrochloride
30007-39-7
C19H21NO2·HCl
2-{[β-(4-Hydroxyphenyl)ethyl]aminomethyl}-1-tetralone hydrochloride
10mg
>98%
331.84

Chemical Identifiers

>98%
331.84
HEAT hydrochloride
C19H21NO2·HCl
2-{[β-(4-Hydroxyphenyl)ethyl]aminomethyl}-1-tetralone hydrochloride

Safety and Handling

Recommended Storage : Desiccate at -20°C