Please login to your online account to display your discounted pricing

HEAT hydrochloride, Tocris

Highly selective α1 antagonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material HEAT hydrochloride
Assay Percent Range >98%
Molecular Formula C19H21NO2·HCl
Formula Weight 331.84
Synonym 2-{[β-(4-Hydroxyphenyl)ethyl]aminomethyl}-1-tetralone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

053550

 
r & d systems
RDS053550
50mg Each for $682.67

053510

 
r & d systems
RDS053510
10mg Each for $164.61
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 331.84
Chemical Name or Material HEAT hydrochloride
Assay Percent Range >98%
Molecular Formula C19H21NO2·HCl
CAS 30007-39-7
Synonym 2-{[β-(4-Hydroxyphenyl)ethyl]aminomethyl}-1-tetralone hydrochloride
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C