Please login to your online account to display your discounted pricing

HDS 029, Tocris

Potent inhibitor of the ErbB receptor family

$136.87 - $290.95

Chemical Identifiers

Chemical Name or Material HDS 029
Assay Percent Range >97%
Molecular Formula C17H11ClFN5O
Formula Weight 355.76
Synonym N-(4-[(-3-Chloro-4-fluorophenyl)amino]pyrido[3, 4-d]pyrimidin-6-yl-2-butynamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

264610

 
r & d systems
264610
10mg Each for $290.95

26461

 
r & d systems
26461
1mg Each for $136.87
Description & Specifications

Specifications

Formula Weight 355.76
Quantity 10mg
CAS 881001-19-0
Chemical Name or Material HDS 029
Assay Percent Range >97%
Molecular Formula C17H11ClFN5O
Synonym N-(4-[(-3-Chloro-4-fluorophenyl)amino]pyrido[3, 4-d]pyrimidin-6-yl-2-butynamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C