Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ HDJ-2/DNAJ Ab-1, Mouse Monoclonal Antibody


Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS225P1ABX

 View more versions of this product

Catalog No. MS225P1ABX

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bacteria
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Non-Human Primate
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Isotype IgG1
Clone KA2A5.6
Regulatory Status RUO
Format Concentrated
Host Species Mouse
Immunogen N-terminal purified recombinant fragment (aa 1-179) of HDJ-2 fusion protein
Antigen HDJ-2/DNAJ Ab-1
Research Discipline Stress Response
Concentration 1mg/mL
Conjugate Unlabeled
Type Primary
Formulation Ab Without BSA/Azide (Frozen)
Classification Monoclonal

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments with Thermo Scientific™ HDJ-2/DNAJ Ab-1, Mouse Monoclonal Antibody.

It is a novel human homolog of E. coli's DNAJ protein from heat shock protein 40 family. Hsp40 is involved in chaperonic protein folding HDJ-2 is cytoplasmic and nuclear membrane associated protein; upon heat shock it migrates and becomes associated with the Golgi complex, nucleolus and nuclear membrane HDJ-2 has a prenylated C-terminus and is not inducible upon heat shock.

Host Species: Mouse

Clone: KA2A5.6

Isotype: IgG1

Species Reactivity: Human, Monkey, Pig, Rabbit, Mouse, Rat, Hamster and Bacteria.

Epitope: aa 1-179

Immunogen: N-terminal purified recombinant fragment (aa 1-179) of HDJ-2 fusion protein

Molecular Weight: 44-47 kDa

Positive Control: MAD109, Raji cells, Tonsil

Cellular Localization: Cytoplasm (ground state) and Golgi complex, Nucleolus, and Nuclear membrane (upon heat shock)

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO