missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hazwik™ Chemical Absorbent Pillows
Surfactant-treated polypropylene designed for absorption of a wide range of chemicals, including hydrofluoric acid.
$86.97 - $213.48
Specifications
Color | Green |
---|---|
Material | Polypropylene |
Type | Hazwik Sorbent Pillow |
For Use With (Application) | Provide extra protection during packaging and transport, absorbs chemicals, acids and bases, spills and leaking fluids in tight spaces |
Catalog Number | Mfr. No. | Dimensions (L x W) | Packaging Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Packaging Quantity | Price | Quantity | ||||
19150274
|
Brady™
HAZ1818 |
45.72 x 45.72 cm (18 x 18 in.) | 16/Cs. |
Case of 16 Each for $213.48
|
|
||||
17-111-164
|
Brady™
HAZ18182 |
45.72 x 45.72 cm (18 x 18 in.) | 8/Cs. |
Case of 8 Each for $86.97
|
|
||||
Description
- Chemically inert — will not react with aggressive fluids
- Color-coded green indicates use with hazardous materials
- Great for absorbing spills and leaking fluids in tight spaces
- Class: chemical
HAZ1818
- Shipping weight: 8.62kg (19 lbs.); 16 per case
- Shipping weight: 5.44kg (12 lbs.); 32 per case
Specifications
Green | |
Polypropylene | |
Hazwik Sorbent Pillow | |
Provide extra protection during packaging and transport, absorbs chemicals, acids and bases, spills and leaking fluids in tight spaces |