Please login to your online account to display your discounted pricing

Hazardous Materials Classification Reference


Hazmat chart is perfect for training or quick reference.

Manufacturer: fisher science education  HMC14PEMD

 View more versions of this product

Catalog No. S47352D

  • / Each
 

Description & Specifications

Specifications

Width (Metric) 25cm
Material Vinyl
Height (Metric) 36cm
Width (English) 10 in.
For Use With (Application) For training or quick reference, graphically explains the NFPA hazard identification coding system
Type Chemical Safety Chart
Height (English) 14 in.