Please login to your online account to display your discounted pricing
Hastings-Type Wide-Angle Corrected Lens Magnifiers
Amplify details with these lenses
$89.45 - $89.45
Specifications
Product Type Hastings Triplet Magnifier
For Use With (Application) To amplify details
Includes Swing away Nickel plated case
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Lens Diopter Length (English) Focal Magnification Price Quantity    
S67131
fisher science education
81-61-75/EM
0.49 in. 56 diopter 0.8 in. 14x Each for $89.45
Description & Specifications
Specifications
Product Type Hastings Triplet Magnifier
For Use With (Application) To amplify details
Includes Swing away Nickel plated case
  • Hastings lens carefully constructed for correct surface and chromatic aberrations
  • Wide-angle viewing available for magnifications of 7X, 10X, and 14X
  • Swing-away, nickle-plated case protects the lens and serves as a handle