missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Harvard Apparatus™ Stronghold™ Lattice Rods

Sturdy rods made to metric specifications

Manufacturer:  Harvard Apparatus™ 532570

 View more versions of this product

Catalog No. 14-679-25



Description

Description

  • Brilliantly polished stainless steel with good surface hardness that will not score from clamp binding screws
  • Hollow, lightweight 13mm (0.5 in.) diameter rods with continuously welded walls 1.6mm (0.16 in.) thick
  • Chemical resistance
  • Red caps

Specifications

Specifications

0.5 in.
Harvard Apparatus™ Stronghold™
150cm
Lattice Rod
1.3cm
59.1 in.
Stainless Steel
0.062 in.
SDS
Documents