missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Harmine, 98+%

$62.92 - $144.34

Chemical Identifiers

CAS 442-51-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00004958
InChI Key BXNJHAXVSOCGBA-UHFFFAOYSA-N
Synonym harmine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterine, leucoharmine, telepathine, yageine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, banisterin
PubChem CID 5280953
ChEBI CHEBI:28121
IUPAC Name 7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
SMILES CC1=NC=CC2=C1NC3=C2C=CC(=C3)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1906803 View Documents Alfa Aesar™
L1906803
1g Each for $144.34
 
AAL19068MD View Documents Alfa Aesar™
L19068MD
250mg Each for $62.92
 
Description

Description

Applications
Harmine, inhibits the Wnt signaling pathway in a cell-specific manner. Down-regulates inflammatory gene expression (TNF-α, IL-1β, iNOS). Reversibly inhibits MAO-A (monoamine oxidase A) but has no effect on MAO-B. Harmine is an inhibitor of Dyrk1A, Dyrk2 and Dyrk3.

Solubility
Soluble in DMSO (100 mM), DMF (~1.5 mg/ml), ethanol (~1.5 mg/ml), and PBS (pH 7.2, ~0.25 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Chemical Identifiers

442-51-3
212.252
BXNJHAXVSOCGBA-UHFFFAOYSA-N
5280953
7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
C13H12N2O
MFCD00004958
harmine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterine, leucoharmine, telepathine, yageine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, banisterin
CHEBI:28121
CC1=NC=CC2=C1NC3=C2C=CC(=C3)OC

Specifications

Odorless
1g
C13H12N2O
178813
harmine, 7-methoxy-1-methyl-9h-pyrido 3,4-b indole, banisterine, leucoharmine, telepathine, yageine, yajeine, 7-methoxy-1-methyl-9h-beta-carboline, 9h-pyrido 3,4-b indole, 7-methoxy-1-methyl, banisterin
CC1=NC=CC2=C1NC3=C2C=CC(=C3)OC
212.252
CHEBI:28121
≥98%
262°C to 266°C
442-51-3
MFCD00004958
14,4616
BXNJHAXVSOCGBA-UHFFFAOYSA-N
7-methoxy-1-methyl-9H-pyrido[3,4-b]indole
5280953
212.25
Harmine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H319
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : UV0175000