Please login to your online account to display your discounted pricing
Hardy Diagnostics Standard Methods Agar
Manufacturer: hardy diagnostics  U295
 View more versions of this product
Catalog No. 02-686-213
  • / Pack of 12 Each
Description & Specifications
Specifications
Quantity 12/Pk.
Media Type Plate count agar/Tryptone glucose yeast (TGY) agar
For Use With (Application) Cultivation and enumeration of microorganisms in dairy products, food, and water