Please login to your online account to display your discounted pricing

Hardness Titrating Solution, 1mL = 1mg CaCO3, Ricca Chemical


Manufacturer: ricca chemical company  RIR34790004F

 View more versions of this product

Catalog No. R34790004F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Chemical Name or Material Hardness Titrating Solution
Shelf Life 12 months
CAS 7791-18-6
Packaging Cubitainer
Name Note 1mL = 1mg CaCO3
CAS 1310-73-2
CAS 6381-92-6
CAS 7732-18-5
Odor Odorless

Specifications

Chemical Name or Material Hardness Titrating Solution
CAS 1310-73-2
CAS 6381-92-6
CAS 7732-18-5
CAS 7791-18-6
Name Note 1mL = 1mg CaCO3
Accessories

12 months