Please login to your online account to display your discounted pricing

Hardness Titrating Solution, 1mL = 1mg CaCO3, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Hardness Titrating Solution
CAS 1310-73-2
CAS 6381-92-6
CAS 7732-18-5
CAS 7791-18-6
Name Note 1mL = 1mg CaCO3
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

R34790004A

 
ricca chemical company
RIR34790004A
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R34790004F

 
ricca chemical company
RIR34790004F
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Chemical Name or Material Hardness Titrating Solution
Shelf Life 12 months
CAS 7791-18-6
Packaging Poly Bottle
Name Note 1mL = 1mg CaCO3
CAS 1310-73-2
CAS 6381-92-6
CAS 7732-18-5
Odor Odorless
Safety and Handling

12 months