Please login to your online account to display your discounted pricing
Hanna™ Instruments pH Buffer Solutions DFS Item
Used for calibration
$27.49 - $29.33
Products
Catalog Number Mfr. No. Price Quantity    
11-603-64
DFS Item
hanna instruments cda inc
HI7010L
Each for $27.56
11-603-68
DFS Item
hanna instruments cda inc
HI7007M
Each for $27.49
11-603-70
DFS Item
hanna instruments cda inc
HI7004M
Each for $29.33
Description & Specifications
  • Available in different pH ranges and quantity
  • Calibrated according to NIST standards
  • With/without certificates of analysis
Safety and Handling
Keep container closed and protected from direct sunlight. Store at room temperature (+15° to +25°C).